Gene Transcripts

Gene Symbol
Acad10
Entrez Gene ID
71985
Gene Name
acyl-Coenzyme A dehydrogenase family, member 10
Gene Aliases
2410021P16Rik
Chromosome Location
Chr. 5: 121621026 - 121660510 on Build GRCm38
UniGene ID
Mm.45423
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028037.4 NP_082313.2 5 792
GenBank mRNA AK050332.1 - 5 996
AK148195.1 - 5 782
BC027825.1 - 5 768 5343012

View Full Product Details