Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001285839.1 | NP_001272768.1 | 4 | 959 | |
GenBank mRNA | AK011265.1 | - | 4 | 907 | |
AK045669.1 | - | 4 | 921 | ||
AK137332.1 | - | 4 | 910 | ||
BC030671.1 | - | 4 | 924 | ||
BC038910.1 | - | 2 | 285 | ||
BC051211.1 | - | 1 | 335 | ||
BC058172.1 | - | 4 | 922 | 5012054 |