Gene Transcripts

Gene Symbol
Dbndd1
Entrez Gene ID
72185
Gene Name
dysbindin (dystrobrevin binding protein 1) domain containing 1
Gene Aliases
2810427I04Rik, D8Ertd590e
Chromosome Location
Chr. 8: 123504718 - 123518041 on Build GRCm38
UniGene ID
Mm.27925
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028146.4 NP_082422.3 3 497
GenBank mRNA AK013173.1 - 3 396
AK028187.1 - 3 434

View Full Product Details