Gene Transcripts

Gene Symbol
Lrpprc
Entrez Gene ID
72416
Gene Name
leucine-rich PPR-motif containing
Gene Aliases
3110001K13Rik, C76645, Gp130, Lrp130, Lsfc
Chromosome Location
Chr. 17: 84705247 - 84790786 on Build GRCm38
UniGene ID
Mm.217027
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028233.2 NP_082509.2 10 1445
GenBank mRNA AK039751.1 - 8 1812
AK138527.1 - 9 1701
BC059862.1 - 10 1179 6417407
BC060199.1 - 10 1217

View Full Product Details