Gene Transcripts

Gene Symbol
Lrpprc
Entrez Gene ID
72416
Gene Name
leucine-rich PPR-motif containing
Gene Aliases
3110001K13Rik, C76645, Gp130, Lrp130, Lsfc
Chromosome Location
Chr. 17: 84705247 - 84790786 on Build GRCm38
UniGene ID
Mm.217027
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028233.2 NP_082509.2 13 1824
GenBank mRNA BC059862.1 - 13 1558
BC060199.1 - 13 1596

View Full Product Details