Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289602.1 | NP_001276531.1 | 2 | 599 | |
NM_028707.4 | NP_082983.3 | 2 | 596 | ||
GenBank mRNA | AK034575.1 | - | 2 | 362 | |
AK045529.1 | - | 2 | 324 | ||
AK053835.1 | - | 2 | 324 | ||
AK149027.1 | - | 2 | 322 | ||
BC062930.1 | - | 1 | 177 | 6855293 | |
BC065053.1 | - | 2 | 361 | 5709231 | |
BC065055.1 | - | 2 | 347 | 5720949 | |
BC066026.1 | - | 2 | 596 | 5721455 | |
BC066036.1 | - | 2 | 362 | 6837266 |