Gene Transcripts

Gene Symbol
Nuak2
Entrez Gene ID
74137
Gene Name
NUAK family, SNF1-like kinase, 2
Gene Aliases
1200013B22Rik, Omphk2, Snark, mKIAA0537
Chromosome Location
Chr. 1: 132316125 - 132333488 on Build GRCm38
UniGene ID
Mm.100666
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195025.1 NP_001181954.1 8 1357
NM_028778.4 NP_083054.1 7 1333
GenBank mRNA AK004737.2 - 7 1321
AK033672.1 - 7 1320
AK034082.1 - 8 1349
BC046833.1 - 8 1332 6816569

View Full Product Details