Gene Transcripts

Gene Symbol
Cep97
Entrez Gene ID
74201
Gene Name
centrosomal protein 97
Gene Aliases
2810403B08Rik, 4932439K18Rik, AA437768, E130116N02Rik, Lrriq2
Chromosome Location
Chr. 16: 55899888 - 55934848 on Build GRCm38
UniGene ID
Mm.178427
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028815.4 NP_083091.1 7 796
GenBank mRNA AK012971.2 - 7 781
AK016556.1 - 7 871
AK046486.1 - 7 1345
AK154885.1 - 7 775
BC067049.1 - 7 796 30363673

View Full Product Details