Gene Transcripts
- Gene Symbol
- Lrrtm1
- Entrez Gene ID
- 74342
- Gene Name
- leucine rich repeat transmembrane neuronal 1
- Gene Aliases
- 4632401D06Rik, AW125451
- Chromosome Location
- Chr. 6: 77242480 - 77259915 on Build GRCm38
- UniGene ID
- Mm.292568
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK053345.1 |
- |
3 |
2537 |
|
AK080836.1 |
- |
3 |
2578 |
|
AK136857.1 |
- |
2 |
2045 |
|
View Full Product Details