Gene Transcripts
- Gene Symbol
- Spata19
- Entrez Gene ID
- 75469
- Gene Name
- spermatogenesis associated 19
- Gene Aliases
- 1700001L23Rik, AV047838, spergen-1
- Chromosome Location
- Chr. 9: 27396821 - 27401711 on Build GRCm38
- UniGene ID
- Mm.45824
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB120716.1 |
- |
2 |
155 |
|
AK005610.1 |
- |
2 |
155 |
|
JX089334.1 |
- |
1 |
327 |
|
View Full Product Details