Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029420.4 | NP_083696.2 | 3 | 707 | |
GenBank mRNA | AK014753.1 | - | 3 | 564 | |
AK049139.1 | - | 3 | 708 | ||
AK053901.1 | - | 2 | 596 | ||
BC145468.1 | - | 2 | 519 | ||
BC145688.1 | - | 3 | 572 | 40130991 |