Gene Transcripts

Gene Symbol
Asb6
Entrez Gene ID
72323
Gene Name
ankyrin repeat and SOCS box-containing 6
Gene Aliases
2510004M11Rik, AA409356
Chromosome Location
Chr. 2: 30823097 - 30828300 on Build GRCm38
UniGene ID
Mm.27656
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133346.2 NP_579924.1 2, 3 398
GenBank mRNA AF398967.1 - 2436
AK010909.1 - 2, 3 379
AK049312.1 - 2, 3 387
AK088890.1 - 2, 3 333
AY665653.1 - 2, 3 284
BC019192.1 - 2, 3 390 5035395
BC085485.1 - 2, 3 355

View Full Product Details