Gene Transcripts

Gene Symbol
Clmp
Entrez Gene ID
71566
Gene Name
CXADR-like membrane protein
Gene Aliases
9030425E11Rik, ACAM, ASP5, AW557819
Chromosome Location
Chr. 9: 40685964 - 40785553 on Build GRCm38
UniGene ID
Mm.280560
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133733.4 NP_598494.2 7 1935
GenBank mRNA AB040490.1 - 7 1853
AK018527.1 - 2 926
AK028983.1 - 7 1905
AK033723.1 - 7 1937
AK033766.1 - 1 42
AK080481.1 - 7 1892
BC026447.1 - 7 1762 4459717

View Full Product Details