Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290662.1 | NP_001277591.1 | 3, 4 | 361 | |
NM_134471.4 | NP_608301.3 | 3, 4 | 402 | ||
GenBank mRNA | AK015046.1 | - | 3, 4 | 372 | |
AK088047.1 | - | 3, 4 | 343 | ||
AK150222.1 | - | 3, 4 | 333 | ||
AK161309.1 | - | 3, 4 | 361 | ||
BC006841.1 | - | 3, 4 | 362 | 3598157 | |
CF163926.1 | - | 3, 4 | 402 |