Gene Transcripts

Gene Symbol
Rasl10a
Entrez Gene ID
75668
Gene Name
RAS-like, family 10, member A
Gene Aliases
2210403B10Rik, AI852688
Chromosome Location
Chr. 11: 5057981 - 5060383 on Build GRCm38
UniGene ID
Mm.374836
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145216.3 NP_660251.1 3 920
GenBank mRNA AK008807.1 - 3 921
AK044011.1 - 3 921
BC107200.1 - 4 895
BC107201.1 - 4 801 40051192

View Full Product Details