Gene Transcripts

Gene Symbol
Oplah
Entrez Gene ID
75475
Gene Name
5-oxoprolinase (ATP-hydrolysing)
Gene Aliases
1700010G02Rik
Chromosome Location
Chr. 15: 76296603 - 76331101 on Build GRCm38
UniGene ID
Mm.322738
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153122.2 NP_694762.1 8 1117
GenBank mRNA AK158391.1 - 8 1159
BC034522.1 - 8 1117 4236444

View Full Product Details