Gene Transcripts
- Gene Symbol
- Wdr27
- Entrez Gene ID
- 71682
- Gene Name
- WD repeat domain 27
- Gene Aliases
- 0610012K18Rik
- Chromosome Location
- Chr. 17: 14784832 - 14943151 on Build GRCm38
- UniGene ID
- Mm.122698
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_175173.3 |
NP_780382.2
|
2, 3 |
229 |
|
GenBank mRNA |
AK077050.1 |
- |
2, 3 |
230 |
|
View Full Product Details