Gene Transcripts

Gene Symbol
Wfdc1
Entrez Gene ID
67866
Gene Name
WAP four-disulfide core domain 1
Gene Aliases
2310058A03Rik, ps20
Chromosome Location
Chr. 8: 119666365 - 119688226 on Build GRCm38
UniGene ID
Mm.87599
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023395.2 NP_075884.1 6 796
GenBank mRNA AK132541.1 - 6 797
AK142707.1 - 6 665
AK161606.1 - 6 662
BC025850.1 - 6 609
BC038122.1 - 6 638
BC132073.1 - 6 725 40130350
BC132469.1 - 6 725 40130746
BC145305.1 - 6 722 9053859

View Full Product Details