Gene Transcripts
- Gene Symbol
- Fam69a
- Entrez Gene ID
- 67266
- Gene Name
- family with sequence similarity 69, member A
- Gene Aliases
- 2900024C23Rik, AI315274
- Chromosome Location
- Chr. 5: 107908042 - 107987077 on Build GRCm38
- UniGene ID
- Mm.277897
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026062.4 |
NP_080338.1
|
3, 4 |
377 |
|
GenBank mRNA |
AK008504.1 |
- |
3, 4 |
320 |
|
AK013580.1 |
- |
3, 4 |
341 |
|
AK038811.1 |
- |
3, 4 |
334 |
|
AK048979.1 |
- |
3, 4 |
334 |
|
AK050305.1 |
- |
3, 4 |
334 |
|
AK053273.1 |
- |
2, 3 |
182 |
|
AK153488.1 |
- |
3, 4 |
332 |
|
BC055842.1 |
- |
3, 4 |
309 |
|
BC061013.1 |
- |
3, 4 |
329 |
|
BC095935.1 |
- |
3, 4 |
311 |
|
BC116683.1 |
- |
3, 4 |
322 |
40106565 |
View Full Product Details