Gene Transcripts
- Gene Symbol
- LANCL1
- Entrez Gene ID
- 10314
- Gene Name
- LanC like 1
- Gene Aliases
- GPR69A, p40
- Chromosome Location
- Chr. 2: 210431249 - 210477618 on Build GRCh38
- UniGene ID
- Hs.13351
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001136574.1 |
NP_001130046.1
|
3, 4 |
282 |
|
NM_001136575.1 |
NP_001130047.1
|
3, 4 |
277 |
|
NM_006055.2 |
NP_006046.1
|
3, 4 |
371 |
|
XM_005246243.2 |
XP_005246300.1
|
3, 4 |
392 |
|
GenBank mRNA |
AK294674.1 |
- |
3, 4 |
309 |
|
AK312381.1 |
- |
3, 4 |
260 |
|
BC028685.2 |
- |
3, 4 |
282 |
4829574 |
CR541777.1 |
- |
2, 3 |
207 |
|
DC321883.1 |
- |
3, 4 |
277 |
|
DC324843.1 |
- |
3, 4 |
372 |
|
Y11395.1 |
- |
3, 4 |
311 |
|
View Full Product Details