Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006516.2 | NP_006507.2 | 2, 3 | 647 | |
GenBank mRNA | AB208987.1 | - | 2, 3 | 367 | |
AK292791.1 | - | 2, 3 | 333 | ||
AK293306.1 | - | 2, 3 | 339 | ||
AK296736.1 | - | 2, 3 | 339 | ||
AK312403.1 | - | 2, 3 | 339 | ||
BC118590.1 | - | 2, 3 | 268 | 40085221 | |
BC121804.1 | - | 2, 3 | 268 | 40085220 | |
K03195.1 | - | 2, 3 | 301 |