Gene Transcripts

Gene Symbol
Tctn3
Entrez Gene ID
67590
Gene Name
tectonic family member 3
Gene Aliases
4930521E07Rik, AI197391, Tect3
Chromosome Location
Chr. 19: 40596446 - 40612281 on Build GRCm38
UniGene ID
Mm.374056
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026260.2 NP_080536.2 10 1213
GenBank mRNA AK015861.1 - 10 1201
AK016982.1 - 10 1199
BC027335.1 - 10 1213 3710046
BC080805.1 - 10 1188 5352578
DQ278871.1 - 10 1230

View Full Product Details