Gene Transcripts

Gene Symbol
Cdca5
Entrez Gene ID
67849
Gene Name
cell division cycle associated 5
Gene Aliases
2610036L13Rik, AL024086, AW536684, C85404
Chromosome Location
Chr. 19: 6085097 - 6091773 on Build GRCm38
UniGene ID
Mm.23526
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026410.3 NP_080686.1 2 143
GenBank mRNA AK010540.1 - 2 110
AK011701.1 - 2 143
AK016305.1 - 2 257
BC052904.1 - 2 141 30065848

View Full Product Details