Gene Transcripts
- Gene Symbol
- Ube2f
- Entrez Gene ID
- 67921
- Gene Name
- ubiquitin-conjugating enzyme E2F (putative)
- Gene Aliases
- 2510010F15Rik, AI851109
- Chromosome Location
- Chr. 1: 91250306 - 91286516 on Build GRCm38
- UniGene ID
- Mm.337238
- Species
- Mus musculus
- Gene Symbol
- Gm5434
- Entrez Gene ID
- 432649
- Gene Name
- ubiquitin-conjugating enzyme E2F (putative) pseudogene
- Gene Aliases
- EG432649
- Chromosome Location
- Chr. 12: 36090379 - 36091829 on Build GRCm38
- UniGene ID
- Mm.347860
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK136183.1 |
- |
1 |
620 |
|
BC085248.1 |
- |
1 |
626 |
|
View Full Product Details