Gene Transcripts
- Gene Symbol
- Hormad1
- Entrez Gene ID
- 67981
- Gene Name
- HORMA domain containing 1
- Gene Aliases
- 4921522K05Rik, Nohma
- Chromosome Location
- Chr. 3: 95559640 - 95587940 on Build GRCm38
- UniGene ID
- Mm.179050
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001289532.1 |
NP_001276461.1
|
12, 13 |
1023 |
|
NM_001289534.1 |
NP_001276463.1
|
12, 13 |
1023 |
|
NM_001289537.1 |
NP_001276466.1
|
12, 13 |
1023 |
|
NM_026489.3 |
NP_080765.1
|
12, 13 |
1014 |
|
GenBank mRNA |
AK014945.1 |
- |
12, 13 |
1022 |
|
AK016743.1 |
- |
12, 13 |
1017 |
|
AK163995.1 |
- |
12, 13 |
1022 |
|
AY626343.1 |
- |
12, 13 |
1022 |
|
AY634284.1 |
- |
12, 13 |
1022 |
|
BC051129.1 |
- |
12, 13 |
1022 |
6706580 |
View Full Product Details