Gene Transcripts

Gene Symbol
Murc
Entrez Gene ID
68016
Gene Name
muscle-related coiled-coil protein
Gene Aliases
2310039E09Rik
Chromosome Location
Chr. 4: 48663514 - 48673492 on Build GRCm38
UniGene ID
Mm.133444
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026509.3 NP_080785.2 2 915
GenBank mRNA AK009691.1 - 2 872
BC089601.1 - 2 915 6788090
EU487254.1 - 2 807

View Full Product Details