Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026532.3 | NP_080808.1 | 5 | 1226 | |
GenBank mRNA | AK028410.1 | - | 5 | 1216 | |
AK188763.1 | - | 1 | 397 | ||
BC003955.1 | - | 5 | 1226 | 3487899 | |
BC086773.1 | - | 5 | 1193 | 6828661 |