Gene Transcripts

Gene Symbol
Dcakd
Entrez Gene ID
68087
Gene Name
dephospho-CoA kinase domain containing
Gene Aliases
3010024O21Rik, 6720485C15Rik, AI849483
Chromosome Location
Chr. 11: 102994047 - 103028239 on Build GRCm38
UniGene ID
Mm.277449
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026551.3 NP_080827.2 3 497
GenBank mRNA AK051022.1 - 3 498
AK078460.1 - 3 430
AK162069.1 - 3 457
AK170762.1 - 3 448
AK190020.1 - 1 194
BC005607.1 - 3 463 3498151
BC023208.1 - 2 411
BC031855.1 - 3 446
BC042685.1 - 2 411
BC045193.1 - 3 451 5368157
BC047340.1 - 2 411

View Full Product Details