Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026558.4 | NP_080834.3 | 2 | 351 | |
GenBank mRNA | AK013129.1 | - | 2 | 182 | |
AK019574.1 | - | 2 | 179 | ||
AK032690.1 | - | 2 | 331 | ||
AK032976.1 | - | 1 | 648 | ||
AK078438.1 | - | 2 | 332 | ||
AK088705.1 | - | 2 | 332 | ||
BC055848.1 | - | 2 | 161 | ||
CJ064072.1 | - | 2 | 352 |