Gene Transcripts

Gene Symbol
Etaa1
Entrez Gene ID
68145
Gene Name
Ewing tumor-associated antigen 1
Gene Aliases
5730466H23Rik, AA672646
Chromosome Location
Chr. 11: 17938749 - 17953875 on Build GRCm38
UniGene ID
Mm.178684
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026576.3 NP_080852.2 5 806
GenBank mRNA AK018594.1 - 1 103
AK019965.1 - 5 807
AK034138.1 - 5 799
BC028916.1 - 5 801 3676092

View Full Product Details