Gene Transcripts

Gene Symbol
Zdhhc3
Entrez Gene ID
69035
Gene Name
zinc finger, DHHC domain containing 3
Gene Aliases
1110020O22Rik, 1810006O10Rik, 2210017C02Rik, DHHC-3, GODZ, Gramp1, Zfp373
Chromosome Location
Chr. 9: 123066802 - 123113205 on Build GRCm38
UniGene ID
Mm.28300
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026917.5 NP_081193.2 7 1550
GenBank mRNA AK018755.1 - 7 1518
AK080541.1 - 7 1517
BC005531.1 - 8 1577
BC018334.1 - 6 1386
BC025126.1 - 7 1536 3669723
BC052464.1 - 7 1457 6834763
BU921579.1 - 1 436

View Full Product Details