Gene Transcripts

Gene Symbol
Cdc16
Entrez Gene ID
69957
Gene Name
CDC16 cell division cycle 16
Gene Aliases
2700071J12Rik, 2810431D22Rik, APC6
Chromosome Location
Chr. 8: 13757646 - 13781938 on Build GRCm38
UniGene ID
Mm.182412
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027276.2 NP_081552.2 17 1651
GenBank mRNA AK013213.1 - 6 585
AK147938.1 - 17 1666
AK167965.1 - 17 1666
AK168978.1 - 17 1638
AK181696.1 - 3 233
AK184248.1 - 3 222
AK188605.1 - 3 227
AK198756.1 - 2 84
AK205790.1 - 3 208
AK214776.1 - 3 180
BC005658.1 - 1 248
BC026606.1 - 17 1651 4976424

View Full Product Details