Gene Transcripts

Gene Symbol
Wdr77
Entrez Gene ID
70465
Gene Name
WD repeat domain 77
Gene Aliases
2610003I18Rik, 2610312E17Rik, C79984, p44/MEP50
Chromosome Location
Chr. 3: 105959498 - 105969760 on Build GRCm38
UniGene ID
Mm.5110
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027432.3 NP_081708.1 10 3389
GenBank mRNA AK050391.1 - 10 3389
AK161812.1 - 9 3847

View Full Product Details