Gene Transcripts
- Gene Symbol
- Tm9sf2
- Entrez Gene ID
- 68059
- Gene Name
- transmembrane 9 superfamily member 2
- Gene Aliases
- 1500001N15Rik, AA536814, D14Ertd64e, P76
- Chromosome Location
- Chr. 14: 122107082 - 122159603 on Build GRCm38
- UniGene ID
- Mm.275191
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_080556.3 |
NP_542123.3
|
4, 5 |
644 |
|
GenBank mRNA |
AK010325.1 |
- |
4, 5 |
601 |
|
AK030340.1 |
- |
4, 5 |
574 |
|
AK048012.1 |
- |
4, 5 |
563 |
|
AK050338.1 |
- |
4, 5 |
591 |
|
AK075720.1 |
- |
4, 5 |
644 |
|
AK088905.1 |
- |
4, 5 |
616 |
|
AK159757.1 |
- |
4, 5 |
589 |
|
AK206314.1 |
- |
1, 2 |
123 |
|
BC003862.1 |
- |
4, 5 |
583 |
3488326 |
View Full Product Details