Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_183270.2 | NP_899093.1 | 2 | 724 | |
GenBank mRNA | AK008883.1 | - | 2 | 690 | |
AK019895.1 | - | 696 | |||
AK088786.1 | - | 2 | 712 | ||
AK161506.1 | - | 725 | |||
BC048574.1 | - | 2 | 665 | ||
BC096605.1 | - | 2 | 691 | 5371531 |