Gene Transcripts
- Gene Symbol
- Hnf4aos
- Entrez Gene ID
- 68314
- Gene Name
- hepatic nuclear factor 4 alpha, opposite strand
- Gene Aliases
- 0610008F07Rik
- Chromosome Location
- Chr. 2: 163493804 - 163541721 on Build GRCm38
- UniGene ID
- Mm.46190
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK002328.1 |
- |
3 |
507 |
|
BC025862.1 |
- |
4 |
702 |
5050923 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
NM_197983.1 |
- |
|
702 |
|
Chr. - - [-] |
View Full Product Details