Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_197987.2 | NP_932104.1 | 24 | 3845 | |
GenBank mRNA | AB093271.1 | - | 24 | 3816 | |
AK029570.1 | - | 10 | 2123 | ||
AK086798.1 | - | 11 | 2221 | ||
AK140822.1 | - | 24 | 3846 | ||
BC022117.1 | - | 19 | 3008 | ||
BC024949.1 | - | 5 | 1168 | ||
BC058678.1 | - | 24 | 3843 | 6834513 | |
BC059070.1 | - | 24 | 3811 | 6825999 | |
BC061474.1 | - | 3 | 846 | ||
BC078416.1 | - | 24 | 3830 |