Gene Transcripts

Gene Symbol
Acp6
Entrez Gene ID
66659
Gene Name
acid phosphatase 6, lysophosphatidic
Gene Aliases
5730559A09Rik, ACPL1, AU022842, mPACPL1
Chromosome Location
Chr. 3: 97158752 - 97176576 on Build GRCm38
UniGene ID
Mm.101368
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019800.4 NP_062774.2 2 631
GenBank mRNA AB030038.1 - 2 624
AF216223.1 - 2 108
AK002783.1 - 2 545
AK077729.1 - 2 619
AK160271.1 - 2 550
AK165356.1 - 2 632
AK171757.1 - 2 546
BC047276.1 - 2 371 6487551

View Full Product Details