Gene Transcripts

Gene Symbol
Mogs
Entrez Gene ID
57377
Gene Name
mannosyl-oligosaccharide glucosidase
Gene Aliases
1810017N02Rik, AI181835, Gcs1
Chromosome Location
Chr. 6: 83115506 - 83118902 on Build GRCm38
UniGene ID
Mm.28188
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020619.2 NP_065644.2 1 383
GenBank mRNA AK144946.1 - 1 384
AK157286.1 - 1 380
BC051949.1 - 1 383 5716189

View Full Product Details