Gene Transcripts

Gene Symbol
Cst10
Entrez Gene ID
58214
Gene Name
cystatin 10 (chondrocytes)
Gene Aliases
DD72
Chromosome Location
Chr. 2: 149405249 - 149410278 on Build GRCm38
UniGene ID
Mm.117117
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021405.2 NP_067380.1 1 344
GenBank mRNA AB036743.1 - 1 344
BC048364.1 - 1 260 4920219

View Full Product Details