Gene Transcripts
- Gene Symbol
- GMEB1
- Entrez Gene ID
- 10691
- Gene Name
- glucocorticoid modulatory element binding protein 1
- Gene Aliases
- P96PIF, PIF96
- Chromosome Location
- Chr. 1: 28668728 - 28715603 on Build GRCh38
- UniGene ID
- Hs.632373
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001319674.1 |
NP_001306603.1
|
3, 4 |
355 |
|
NM_006582.3 |
NP_006573.2
|
3, 4 |
385 |
|
NM_024482.2 |
NP_077808.1
|
3, 4 |
352 |
|
XM_011540518.2 |
XP_011538820.1
|
3, 4 |
488 |
|
XM_011540519.2 |
XP_011538821.1
|
3, 4 |
328 |
|
XM_011540521.2 |
XP_011538823.1
|
3, 4 |
457 |
|
XM_011540522.2 |
XP_011538824.1
|
3, 4 |
297 |
|
XM_017000087.1 |
XP_016855576.1
|
3, 4 |
503 |
|
GenBank mRNA |
AF099013.1 |
- |
2, 3 |
249 |
|
AF173868.1 |
- |
3, 4 |
280 |
|
AF203694.1 |
- |
3, 4 |
279 |
|
AK000892.1 |
- |
3, 4 |
351 |
|
AK023126.1 |
- |
3, 4 |
272 |
|
AK315195.1 |
- |
3, 4 |
331 |
|
AK315436.1 |
- |
3, 4 |
289 |
|
BC001473.2 |
- |
3, 4 |
302 |
3138481 |
DA919003.1 |
- |
3, 4 |
382 |
|
View Full Product Details