Gene Transcripts

Gene Symbol
Treh
Entrez Gene ID
58866
Gene Name
trehalase (brush-border membrane glycoprotein)
Gene Aliases
2210412M19Rik
Chromosome Location
Chr. 9: 44673233 - 44686305 on Build GRCm38
UniGene ID
Mm.45380
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277847.1 NP_001264776.1 7 737
NM_001277851.1 NP_001264780.1 7 741
NM_021481.3 NP_067456.1 8 842
GenBank mRNA AF136944.1 - 8 837
AK002566.1 - 7 740
AK008912.1 - 8 827
BC019214.1 - 7 732 5064708

View Full Product Details