Gene Transcripts

Gene Symbol
Pxmp4
Entrez Gene ID
59038
Gene Name
peroxisomal membrane protein 4
Gene Aliases
3010018P03Rik, Pmp24
Chromosome Location
Chr. 2: 154587044 - 154603673 on Build GRCm38
UniGene ID
Mm.24673
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021534.3 NP_067509.1 4 1238
GenBank mRNA AK034672.1 - 105
AK050647.1 - 4 1239
AK156743.1 - 1238
BC025796.1 - 4 1217 5100832

View Full Product Details