Gene Transcripts

Gene Symbol
Nek6
Entrez Gene ID
59126
Gene Name
NIMA (never in mitosis gene a)-related expressed kinase 6
Gene Aliases
1300007C09Rik
Chromosome Location
Chr. 2: 38511697 - 38587490 on Build GRCm38
UniGene ID
Mm.143818
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159631.1 NP_001153103.1 10 1014
NM_021606.3 NP_067619.1 10 1007
GenBank mRNA AF218847.1 - 10 1009
AK004925.1 - 10 984
AK011519.1 - 10 993
AK018168.1 - 10 996
AK029266.1 - 10 1012
AK075717.1 - 10 981
AK075836.1 - 10 993
AK086700.1 - 1012
AK089919.1 - 10 968
AK170757.1 - 10 1014
AK185671.1 - 3 275
BC019524.1 - 10 1076 4209097

View Full Product Details