Gene Transcripts

Gene Symbol
Fn3k
Entrez Gene ID
63828
Gene Name
fructosamine 3 kinase
Gene Aliases
2310074G21Rik
Chromosome Location
Chr. 11: 121434809 - 121450491 on Build GRCm38
UniGene ID
Mm.266448
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022014.4 NP_071297.1 6 725
GenBank mRNA AJ404616.1 - 6 711
AK010151.1 - 6 719
BU571242.1 - 4 354

View Full Product Details