Gene Transcripts

Gene Symbol
Zmat2
Entrez Gene ID
66492
Gene Name
zinc finger, matrin type 2
Gene Aliases
2610510D14Rik, 2900082I05Rik
Chromosome Location
Chr. 18: 36793923 - 36799660 on Build GRCm38
UniGene ID
Mm.251155
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025594.3 NP_079870.1 6 1564
GenBank mRNA BC024881.1 - 6 1574

View Full Product Details