Gene Transcripts

Gene Symbol
Spryd7
Entrez Gene ID
66674
Gene Name
SPRY domain containing 7
Gene Aliases
6330409N04Rik, Clld6
Chromosome Location
Chr. 14: 61533879 - 61556886 on Build GRCm38
UniGene ID
Mm.282706
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025697.3 NP_079973.1 3 535
GenBank mRNA AK018044.1 - 3 413
AK018153.1 - 3 536
AK018290.1 - 3 385
AK137685.1 - 384
AK150789.1 - 3 410
AK167306.1 - 3 377
AK169059.1 - 3 384
BC037015.1 - 3 369 5344409

View Full Product Details