Gene Transcripts

Gene Symbol
Dynlt3
Entrez Gene ID
67117
Gene Name
dynein light chain Tctex-type 3
Gene Aliases
2310075M16Rik, AU042796, Tcte1l
Chromosome Location
Chr. X: 9654270 - 9662983 on Build GRCm38
UniGene ID
Mm.29150
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025975.5 NP_080251.3 4 314
GenBank mRNA AK010186.1 - 4 248
AK018842.1 - 4 292
AK088277.1 - 4 315
BC030032.1 - 4 241 1332100

View Full Product Details