Gene Transcripts
- Gene Symbol
- Ghitm
- Entrez Gene ID
- 66092
- Gene Name
- growth hormone inducible transmembrane protein
- Gene Aliases
- 1010001P14Rik, C77840, MICS1, PTD010, Tmbim5
- Chromosome Location
- Chr. 14: 37120444 - 37135327 on Build GRCm38
- UniGene ID
- Mm.182912
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199122.1 |
NP_001186051.1
|
4, 5 |
550 |
|
NM_078478.5 |
NP_510963.1
|
4, 5 |
503 |
|
GenBank mRNA |
AF412297.1 |
- |
4, 5 |
447 |
|
AK005636.1 |
- |
4, 5 |
447 |
|
AK093887.1 |
- |
4, 5 |
479 |
|
AK134592.1 |
- |
4, 5 |
433 |
|
AK146433.1 |
- |
4, 5 |
410 |
|
AK146853.1 |
- |
4, 5 |
466 |
|
AK151488.1 |
- |
4, 5 |
480 |
|
AK158904.1 |
- |
4, 5 |
493 |
|
AK159695.1 |
- |
4, 5 |
477 |
|
AK159724.1 |
- |
4, 5 |
477 |
|
AK164122.1 |
- |
4, 5 |
448 |
|
AK167970.1 |
- |
4, 5 |
474 |
|
AK168022.1 |
- |
4, 5 |
480 |
|
AK168678.1 |
- |
4, 5 |
551 |
|
AK168800.1 |
- |
4, 5 |
459 |
|
BC008622.1 |
- |
4, 5 |
481 |
3588758 |
BC010224.1 |
- |
4, 5 |
483 |
3500685 |
View Full Product Details