Gene Transcripts

Gene Symbol
Lpar6
Entrez Gene ID
67168
Gene Name
lysophosphatidic acid receptor 6
Gene Aliases
2610302I02Rik, P2ry5, P2y5
Chromosome Location
Chr. 14: 73237891 - 73240358 on Build GRCm38
UniGene ID
Mm.390681
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175116.4 NP_780325.1 1 879
GenBank mRNA AK008952.1 - 1 755
AK011967.1 - 1 771
AK032901.1 - 1 876
AK169602.1 - 1 768
BC047931.1 - 1 755 5251236
BC057175.1 - 1 742
BC069991.1 - 1 728 30244233
BC145940.1 - 1 505 40131243
BC145942.1 - 1 505 40131245

View Full Product Details